The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005761.2 | NP_005752.1 | 17 - 18 | 3339 | 141 |
XM_011537730.2 | XP_011536032.1 | 17 - 18 | 3366 | 141 | |
XM_017018671.1 | XP_016874160.1 | 17 - 18 | 3366 | 141 | |
XM_017018672.1 | XP_016874161.1 | 17 - 18 | 3366 | 141 | |
GenBank mRNA | AB208934.1 | - | 16 - 17 | 2020 | 141 |
AF030339.1 | - | 17 - 18 | 3339 | 141 | |
AK295226.1 | - | 2 - 3 | 468 | 141 | |
AK308985.1 | - | 3 - 4 | 272 | 141 |