Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUX2
Entrez Gene ID
23316
Gene Name
cut like homeobox 2
Gene Aliases
CDP2, CUTL2
Chromosome Location
Chr.12: 111034024 - 111350556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015267.3 NP_056082.2 5 - 6 588 129
XM_011538061.2 XP_011536363.1 5 - 6 2098 129
XM_011538069.2 XP_011536371.1 5 - 6 437 129
XM_011538070.2 XP_011536372.1 5 - 6 1347 129
XM_017019080.1 XP_016874569.1 6 - 7 1430 129
XM_017019081.1 XP_016874570.1 6 - 7 493 129
GenBank mRNA AB006631.2 - 5 - 6 587 129
AF234996.1 - 6 - 7 751 129
AF271236.1 - 5 - 6 498 129
BC151245.1 - 5 - 6 598 129

View Full Product Details