Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C7orf43
Entrez Gene ID
55262
Gene Name
chromosome 7 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.7: 100154420 - 100158721 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK001787.1 - 5 - 6 693 78

View Full Product Details