Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG5
Entrez Gene ID
9231
Gene Name
discs large MAGUK scaffold protein 5
Gene Aliases
LP-DLG, P-DLG5, PDLG
Chromosome Location
Chr.10: 77790787 - 77926590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004747.3 NP_004738.3 27 - 28 5230 71
XM_005270276.4 XP_005270333.1 27 - 28 5184 71
XM_006718056.3 XP_006718119.1 26 - 27 4173 71
XM_011540341.2 XP_011538643.1 27 - 28 5354 71
XM_011540342.1 XP_011538644.1 27 - 28 4932 71
XM_011540344.2 XP_011538646.1 26 - 27 4839 71
XM_011540345.1 XP_011538647.1 26 - 27 4964 71
XM_011540347.2 XP_011538649.1 23 - 24 4410 71
XM_017016913.1 XP_016872402.1 27 - 28 4893 71
XM_017016914.1 XP_016872403.1 27 - 28 5223 71
XM_017016915.1 XP_016872404.1 23 - 24 5094 71
GenBank mRNA AB011155.2 - 27 - 28 5260 71
AB091676.1 - 25 - 26 4695 71
AK122931.1 - 28 - 29 5229 71
AK314771.1 - 11 - 12 1539 71
BC073996.1 - 13 - 14 1940 71
BC117695.2 - 17 - 18 4988 71
BC146794.1 - 27 - 28 5260 71
U61843.1 - 12 - 13 1788 71

View Full Product Details