The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 24 - 25 | 2793 | 143 |
GenBank mRNA | AB023215.1 | - | 22 - 23 | 2353 | 143 |
AK309748.1 | - | 25 - 26 | 2775 | 143 | |
AL136808.1 | - | 10 - 11 | 1241 | 143 | |
AY237126.1 | - | 24 - 25 | 2723 | 143 | |
BC016625.1 | - | 11 - 12 | 1374 | 143 | |
BC136472.1 | - | 24 - 25 | 2683 | 143 | |
BC136473.1 | - | 24 - 25 | 2683 | 143 | |
BX647530.1 | - | 17 - 18 | 1862 | 143 | |
CR533554.1 | - | 9 - 10 | 1171 | 143 |