The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012093.3 | NP_036225.2 | 11 - 12 | 2262 | 113 |
NM_174858.2 | NP_777283.1 | 11 - 12 | 1651 | 113 | |
XM_005270739.4 | XP_005270796.1 | 11 - 12 | 1498 | 113 | |
XM_006710572.3 | XP_006710635.1 | 11 - 12 | 1429 | 113 | |
XM_017001008.1 | XP_016856497.1 | 11 - 12 | 1446 | 113 | |
XM_017001009.1 | XP_016856498.1 | 11 - 12 | 2154 | 113 | |
XM_017001010.1 | XP_016856499.1 | 11 - 12 | 2481 | 113 | |
XM_017001011.1 | XP_016856500.1 | 11 - 12 | 1264 | 113 | |
GenBank mRNA | AF062595.1 | - | 3 - 4 | 269 | 113 |
AF445193.1 | - | 11 - 12 | 1651 | 113 | |
AY171600.2 | - | 11 - 12 | 2267 | 113 | |
BC012467.2 | - | 6 - 7 | 614 | 113 | |
BC033896.1 | - | 11 - 12 | 2267 | 113 | |
BC036666.1 | - | 11 - 12 | 1577 | 113 | |
CR541890.1 | - | 2 - 3 | 219 | 113 |