The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168398.1 | NP_001161870.1 | 2 - 3 | 266 | 126 |
NM_006416.4 | NP_006407.1 | 2 - 3 | 266 | 126 | |
GenBank mRNA | AJ810302.1 | - | 2 - 3 | 202 | 126 |
AJ810303.1 | - | 2 - 3 | 202 | 126 | |
AJ851888.1 | - | 2 - 3 | 187 | 126 | |
AJ851889.1 | - | 2 - 3 | 187 | 126 | |
AK299432.1 | - | 2 - 3 | 214 | 126 | |
AL122119.1 | - | 2 - 3 | 262 | 126 | |
BC017807.1 | - | 2 - 3 | 214 | 126 | |
D87969.1 | - | 2 - 3 | 214 | 126 | |
DA210713.1 | - | 2 - 3 | 272 | 126 | |
DB095595.1 | - | 2 - 3 | 266 | 126 |