Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDN1
Entrez Gene ID
23195
Gene Name
midasin AAA ATPase 1
Gene Aliases
Rea1
Chromosome Location
Chr.6: 89642775 - 89828021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014611.2 NP_055426.1 27 - 28 4086 63
XM_011535635.2 XP_011533937.1 27 - 28 3891 63
XM_011535636.2 XP_011533938.1 27 - 28 4056 63
GenBank mRNA AF503925.1 - 27 - 28 4015 63

View Full Product Details