Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2J2
Entrez Gene ID
1573
Gene Name
cytochrome P450 family 2 subfamily J member 2
Gene Aliases
CPJ2, CYPIIJ2
Chromosome Location
Chr.1: 59893308 - 59926798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_134982.1 7 - 8 1240 64
GenBank mRNA AK055088.1 - 5 - 6 1743 64

View Full Product Details