The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007187.3 | NP_009118.1 | 8 - 9 | 858 | 99 |
XM_005266245.2 | XP_005266302.1 | 8 - 9 | 875 | 99 | |
XM_017020380.1 | XP_016875869.1 | 7 - 8 | 788 | 99 | |
GenBank mRNA | AF071185.1 | - | 8 - 9 | 894 | 99 |
AK000979.1 | - | 8 - 9 | 910 | 100 | |
AK001557.1 | - | 8 - 9 | 893 | 99 | |
AK297536.1 | - | 7 - 8 | 796 | 99 | |
AK314525.1 | - | 8 - 9 | 895 | 99 | |
BC104879.1 | - | 8 - 9 | 802 | 99 | |
BC108310.1 | - | 8 - 9 | 859 | 99 |