The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147193.2 | NP_671726.2 | 8 - 9 | 2105 | 94 |
XM_017000408.1 | XP_016855897.1 | 9 - 10 | 2172 | 94 | |
XM_017000409.1 | XP_016855898.1 | 9 - 10 | 2160 | 94 | |
XM_017000410.1 | XP_016855899.1 | 9 - 10 | 2286 | 94 | |
GenBank mRNA | AK090634.1 | - | 8 - 9 | 2104 | 94 |
AK093474.1 | - | 6 - 7 | 1474 | 94 | |
BC101799.1 | - | 6 - 7 | 1611 | 94 | |
BC104911.1 | - | 6 - 7 | 1611 | 94 |