Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
Chromosome Location
Chr.18: 23992773 - 24135952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135993.1 NP_001129465.1 6 - 7 1435 140
NM_153211.3 NP_694943.2 6 - 7 1082 140
XM_005258198.4 XP_005258255.1 8 - 9 1469 140
XM_011525814.2 XP_011524116.1 6 - 7 1623 140
XM_017025550.1 XP_016881039.1 8 - 9 1563 140
GenBank mRNA AK091080.1 - 6 - 7 1082 140
BC032684.1 - 6 - 7 884 140
BC121034.2 - 6 - 7 965 140
BC121035.2 - 6 - 7 965 140

View Full Product Details