The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207354.2 | NP_997237.2 | 7 - 8 | 968 | 76 |
NR_030767.1 | 8 - 9 | 1051 | 76 | ||
XM_011544985.1 | XP_011543287.1 | 7 - 8 | 911 | 76 | |
XM_011544986.1 | XP_011543288.1 | 8 - 9 | 1051 | 76 | |
XM_011544987.2 | XP_011543289.1 | 7 - 8 | 885 | 76 | |
XM_011544988.2 | XP_011543290.1 | 2 - 3 | 231 | 76 | |
GenBank mRNA | AK096277.1 | - | 7 - 8 | 881 | 76 |
AK124721.1 | - | 8 - 9 | 1048 | 76 | |
AK126438.1 | - | 6 - 7 | 965 | 76 | |
BC110419.1 | - | 7 - 8 | 941 | 76 | |
BC121024.1 | - | 7 - 8 | 842 | 76 | |
BC121025.2 | - | 7 - 8 | 842 | 76 |