The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243733.1 | NP_001230662.1 | 3 - 4 | 548 | 52 |
NM_003377.4 | NP_003368.1 | 3 - 4 | 548 | 52 | |
GenBank mRNA | BC008818.2 | - | 3 - 4 | 338 | 52 |
U43368.1 | - | 3 - 4 | 300 | 52 | |
U43369.1 | - | 3 - 4 | 298 | 52 | |
U48801.1 | - | 3 - 4 | 301 | 52 | |
U52819.1 | - | 3 - 4 | 302 | 52 |