Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr.10: 109864766 - 109923553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167604.1 NP_001161076.1 6 - 7 625 81
NM_001324128.1 NP_001311057.1 4 - 5 472 81
NM_001324131.1 NP_001311060.1 7 - 8 720 81
NM_001324132.1 NP_001311061.1 5 - 6 536 81
NM_001324133.1 NP_001311062.1 6 - 7 625 81
NM_001324134.1 NP_001311063.1 4 - 5 456 81
NM_001324135.1 NP_001311064.1 6 - 7 625 81
NM_001324136.1 NP_001311065.1 6 - 7 840 81
NM_020383.3 NP_065116.3 6 - 7 625 81
XM_011540134.1 XP_011538436.1 6 - 7 619 81
XM_017016614.1 XP_016872103.1 7 - 8 608 81
XM_017016615.1 XP_016872104.1 5 - 6 492 81
GenBank mRNA AF195530.1 - 5 - 6 452 81
AF272981.1 - 4 - 5 376 81
AK093693.1 - 4 - 5 430 81
AK223513.1 - 5 - 6 436 81
AK304367.1 - 6 - 7 518 81
AL833411.1 - 6 - 7 531 81
BC005126.2 - 5 - 6 414 81
BC007579.1 - 6 - 7 516 81
BC013417.2 - 2 - 3 126 81
BX537682.1 - 3 - 4 281 81
CR456922.1 - 4 - 5 376 81
X95762.1 - 4 - 5 376 81

View Full Product Details