Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD11
Entrez Gene ID
22984
Gene Name
programmed cell death 11
Gene Aliases
ALG-4, ALG4, NFBP, RRP5
Chromosome Location
Chr.10: 103396615 - 103446295 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014976.1 NP_055791.1 33 - 34 5212 64
XM_005269647.3 XP_005269704.1 33 - 34 5255 64
XM_011539538.2 XP_011537840.1 33 - 34 5203 64
XM_011539539.2 XP_011537841.1 33 - 34 5200 64
XM_011539540.1 XP_011537842.1 32 - 33 5069 64
GenBank mRNA D80007.1 - 33 - 34 5165 64

View Full Product Details