The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014976.1 | NP_055791.1 | 4 - 5 | 482 | 72 |
XM_005269647.3 | XP_005269704.1 | 4 - 5 | 522 | 72 | |
XM_011539538.2 | XP_011537840.1 | 4 - 5 | 470 | 72 | |
XM_011539539.2 | XP_011537841.1 | 4 - 5 | 470 | 72 | |
XM_011539540.1 | XP_011537842.1 | 3 - 4 | 336 | 72 | |
GenBank mRNA | AK294971.1 | - | 4 - 5 | 511 | 72 |
AK307418.1 | - | 4 - 5 | 495 | 72 | |
BC049838.1 | - | 4 - 5 | 483 | 72 | |
BC064486.1 | - | 4 - 5 | 475 | 72 | |
BC080560.1 | - | 4 - 5 | 475 | 72 | |
BC111040.1 | - | 4 - 5 | 437 | 72 | |
D80007.1 | - | 4 - 5 | 435 | 72 |