The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020232.4 | NP_064617.2 | 1 - 2 | 178 | 99 |
GenBank mRNA | AF276707.1 | - | 1 - 2 | 738 | 99 |
AK057005.1 | - | 1 - 2 | 110 | 99 | |
AK225882.1 | - | 1 - 2 | 145 | 99 | |
BC013356.2 | - | 1 - 2 | 101 | 99 | |
BF668035.1 | - | 1 - 2 | 179 | 99 |