The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020232.4 | NP_064617.2 | 5 - 6 | 706 | 78 |
NM_147163.1 | NP_671692.1 | 5 - 6 | 564 | 78 | |
GenBank mRNA | AF068295.1 | - | 5 - 6 | 564 | 78 |
AF168712.1 | - | 5 - 6 | 638 | 78 | |
AF276707.1 | - | 5 - 6 | 1266 | 78 | |
AK025410.1 | - | 1 - 2 | 52 | 78 | |
AK057005.1 | - | 5 - 6 | 638 | 78 | |
AK225882.1 | - | 5 - 6 | 673 | 78 | |
BC013356.2 | - | 5 - 6 | 629 | 78 |