The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033274.4 | NP_150377.1 | 12 - 13 | 1439 | 76 |
XM_005266003.2 | XP_005266060.1 | 12 - 13 | 1391 | 76 | |
XM_011534682.2 | XP_011532984.1 | 5 - 6 | 2195 | 76 | |
XM_017010009.1 | XP_016865498.1 | 5 - 6 | 656 | 76 | |
GenBank mRNA | AF134707.1 | - | 9 - 10 | 1020 | 76 |
AF311317.1 | - | 12 - 13 | 1348 | 76 | |
AF326918.1 | - | 12 - 13 | 1360 | 76 | |
AK091464.1 | - | 12 - 13 | 1401 | 76 | |
AK308182.1 | - | 12 - 13 | 1391 | 76 | |
Y13786.2 | - | 12 - 13 | 1389 | 76 |