The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145114.1 | NP_001138586.1 | 10 - 11 | 1375 | 70 |
NM_001284337.1 | NP_001271266.1 | 9 - 10 | 1258 | 70 | |
NM_015179.3 | NP_055994.2 | 12 - 13 | 1558 | 70 | |
XM_011539555.1 | XP_011537857.1 | 11 - 12 | 1427 | 70 | |
XM_011539556.2 | XP_011537858.1 | 12 - 13 | 1528 | 70 | |
GenBank mRNA | AB014590.1 | - | 9 - 10 | 1171 | 70 |
AK000238.1 | - | 9 - 10 | 968 | 70 | |
AK021460.1 | - | - | 57 | 70 | |
AK022496.1 | - | 12 - 13 | 1529 | 70 | |
AK131283.1 | - | 6 - 7 | 634 | 70 | |
AK296315.1 | - | 10 - 11 | 1239 | 70 | |
BC012745.2 | - | 12 - 13 | 1496 | 70 |