The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145114.1 | NP_001138586.1 | 27 - 28 | 3470 | 64 |
NM_001284337.1 | NP_001271266.1 | 26 - 27 | 3353 | 64 | |
NM_015179.3 | NP_055994.2 | 29 - 30 | 3653 | 64 | |
XM_011539555.1 | XP_011537857.1 | 28 - 29 | 3522 | 64 | |
GenBank mRNA | AB014590.1 | - | 26 - 27 | 3266 | 64 |
AK021460.1 | - | - | 824 | 64 | |
AK022496.1 | - | 29 - 30 | 3624 | 64 | |
AK056614.1 | - | 4 - 5 | 803 | 64 | |
AK131283.1 | - | 23 - 24 | 2729 | 64 | |
AK296315.1 | - | 27 - 28 | 3334 | 64 | |
AL832924.1 | - | 12 - 13 | 2499 | 64 | |
BC002621.2 | - | 2 - 3 | 233 | 64 | |
BC012745.2 | - | 29 - 30 | 3591 | 64 |