Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CHD4
- Entrez Gene ID
-
1108
- Gene Name
- chromodomain helicase DNA binding protein 4
- Gene Aliases
-
CHD-4, Mi-2b, Mi2-BETA
- Chromosome Location
-
Chr.12: 6570082 - 6607439 on Build GRCh38
View Full Product Details