Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TMEM8B
- Entrez Gene ID
-
51754
- Gene Name
- transmembrane protein 8B
- Gene Aliases
-
C9orf127, NAG-5, NAG5, NGX6
- Chromosome Location
-
Chr.9: 35825817 - 35865515 on Build GRCh38
View Full Product Details