Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
Chromosome Location
Chr.9: 35825817 - 35865515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016446.3 NP_057530.2 1 - 2 169 126
GenBank mRNA AF188239.1 - 1 - 2 130 126

View Full Product Details