The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002918.4 | NP_002909.4 | 15 - 16 | 2392 | 88 |
XM_011528165.2 | XP_011526467.1 | 16 - 17 | 2722 | 88 | |
XM_011528166.2 | XP_011526468.1 | 16 - 17 | 2658 | 88 | |
XM_011528167.2 | XP_011526469.1 | 16 - 17 | 3267 | 88 | |
XM_011528168.2 | XP_011526470.1 | 16 - 17 | 2428 | 88 | |
XM_011528169.2 | XP_011526471.1 | 15 - 16 | 2318 | 88 | |
XM_011528170.2 | XP_011526472.1 | 15 - 16 | 2896 | 88 | |
GenBank mRNA | A20498.1 | - | 15 - 16 | 2213 | 88 |
BC049826.1 | - | 15 - 16 | 2353 | 88 | |
X58964.1 | - | 15 - 16 | 2213 | 88 |