The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271044.2 | NP_001257973.1 | 8 - 9 | 1293 | 107 |
NM_002501.3 | NP_002492.2 | 8 - 9 | 1324 | 107 | |
XM_005259919.3 | XP_005259976.1 | 8 - 9 | 1448 | 107 | |
XM_006722760.2 | XP_006722823.1 | 7 - 8 | 1325 | 107 | |
XM_017026837.1 | XP_016882326.1 | 7 - 8 | 1459 | 107 | |
GenBank mRNA | AK297261.1 | - | - | 1292 | 107 |
BC117113.1 | - | 7 - 8 | 1287 | 107 | |
BC117115.1 | - | 7 - 8 | 1287 | 107 | |
BT019732.1 | - | 8 - 9 | 1256 | 107 | |
BX537794.1 | - | 7 - 8 | 1207 | 107 | |
L31881.1 | - | 7 - 8 | 1292 | 107 | |
U18759.1 | - | 8 - 9 | 1273 | 107 |