The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 11 - 12 | 1512 | 129 |
XM_005263236.2 | XP_005263293.1 | 11 - 12 | 1567 | 129 | |
GenBank mRNA | AF327722.1 | - | 11 - 12 | 1535 | 129 |
AJ314788.1 | - | 11 - 12 | 1464 | 129 | |
AK023387.1 | - | 11 - 12 | 1233 | 129 | |
AK023402.1 | - | 11 - 12 | 1535 | 129 | |
AK304417.1 | - | 10 - 11 | 1193 | 129 | |
AK314626.1 | - | 11 - 12 | 1567 | 129 | |
AY039242.1 | - | 11 - 12 | 1537 | 129 | |
BC032642.1 | - | 11 - 12 | 1537 | 129 | |
BC039818.1 | - | 11 - 12 | 1527 | 129 | |
BC093928.1 | - | 11 - 12 | 1321 | 129 | |
BC104806.1 | - | 11 - 12 | 1321 | 129 |