The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006946.2 | NP_008877.1 | 17 - 18 | 3844 | 61 |
XM_005274192.4 | XP_005274249.1 | 18 - 19 | 4506 | 61 | |
XM_005274193.3 | XP_005274250.1 | 17 - 18 | 4280 | 61 | |
XM_006718669.3 | XP_006718732.1 | 16 - 17 | 6193 | 61 | |
XM_006718671.3 | XP_006718734.1 | 18 - 19 | 3906 | 61 | |
XM_011545216.2 | XP_011543518.1 | 16 - 17 | 6203 | 61 | |
XM_011545217.2 | XP_011543519.1 | 16 - 17 | 6203 | 61 | |
XM_017018174.1 | XP_016873663.1 | 18 - 19 | 4598 | 61 | |
XM_017018175.1 | XP_016873664.1 | 18 - 19 | 4227 | 61 | |
XM_017018176.1 | XP_016873665.1 | 18 - 19 | 3947 | 61 | |
XM_017018177.1 | XP_016873666.1 | 17 - 18 | 4136 | 61 | |
XM_017018178.1 | XP_016873667.1 | 17 - 18 | 3819 | 61 | |
GenBank mRNA | AB008567.1 | - | 17 - 18 | 3844 | 61 |
BC131819.1 | - | 1 - 2 | 106 | 61 |