The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006946.2 | NP_008877.1 | 22 - 23 | 4667 | 60 |
XM_005274192.4 | XP_005274249.1 | 23 - 24 | 5329 | 60 | |
XM_005274193.3 | XP_005274250.1 | 22 - 23 | 5103 | 60 | |
XM_006718669.3 | XP_006718732.1 | 21 - 22 | 7016 | 60 | |
XM_006718671.3 | XP_006718734.1 | 23 - 24 | 4729 | 60 | |
XM_017018174.1 | XP_016873663.1 | 23 - 24 | 5421 | 60 | |
XM_017018175.1 | XP_016873664.1 | 23 - 24 | 5050 | 60 | |
XM_017018176.1 | XP_016873665.1 | 23 - 24 | 4770 | 60 | |
XM_017018177.1 | XP_016873666.1 | 22 - 23 | 4959 | 60 | |
XM_017018178.1 | XP_016873667.1 | 22 - 23 | 4642 | 60 | |
GenBank mRNA | AB008567.1 | - | 22 - 23 | 4667 | 60 |
BC131819.1 | - | 6 - 7 | 929 | 60 |