Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
Chromosome Location
Chr.5: 157477304 - 157575823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC024214.2 - 11 - 12 1414 116
BC033132.1 - 11 - 12 1414 116

View Full Product Details