Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
Chromosome Location
Chr.5: 157477304 - 157575823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033274.4 NP_150377.1 1 - 2 218 131
XM_005266003.2 XP_005266060.1 1 - 2 170 131
GenBank mRNA AF311317.1 - 1 - 2 127 131
AK308182.1 - 1 - 2 170 131
HY088077.1 - 1 - 2 218 131
Y13786.2 - 1 - 2 168 131

View Full Product Details