The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105538.1 | NP_001099008.1 | 21 - 22 | 3086 | 77 |
NM_014520.3 | NP_055335.2 | 21 - 22 | 3086 | 77 | |
XM_011523616.2 | XP_011521918.1 | 21 - 22 | 3078 | 77 | |
GenBank mRNA | AF147709.1 | - | 21 - 22 | 3049 | 77 |
AK293458.1 | - | 3 - 4 | 592 | 77 | |
AL133098.1 | - | 3 - 4 | 403 | 77 | |
AL136595.1 | - | 20 - 21 | 2794 | 77 | |
AY093673.1 | - | 2 - 3 | 518 | 77 | |
BC000641.2 | - | 5 - 6 | 738 | 77 | |
BC050546.1 | - | 21 - 22 | 3029 | 77 |