The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207013.1 | NP_001193942.1 | 10 - 11 | 1405 | 128 |
NM_080422.2 | NP_536347.1 | 9 - 10 | 1336 | 128 | |
XM_017025888.1 | XP_016881377.1 | 8 - 9 | 1177 | 128 | |
GenBank mRNA | AK225706.1 | - | 10 - 11 | 1284 | 128 |
AK290649.1 | - | 9 - 10 | 1218 | 128 | |
BC008244.1 | - | 9 - 10 | 1179 | 128 | |
M80737.1 | - | 1 - 2 | 95 | 128 | |
M81478.1 | - | 1 - 2 | 95 | 128 |