The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164814.1 | NP_001158286.1 | 9 - 10 | 2769 | 70 |
NM_001164815.1 | NP_001158287.1 | 8 - 9 | 2649 | 70 | |
NM_001164816.1 | NP_001158288.1 | 2 - 3 | 288 | 70 | |
NM_001164817.1 | NP_001158289.1 | 3 - 4 | 350 | 70 | |
NM_014977.3 | NP_055792.1 | 9 - 10 | 2769 | 70 | |
XM_005267415.3 | XP_005267472.1 | 9 - 10 | 2573 | 70 | |
XM_005267416.3 | XP_005267473.1 | 9 - 10 | 2573 | 70 | |
XM_005267418.1 | XP_005267475.1 | 2 - 3 | 585 | 70 | |
GenBank mRNA | AB014570.2 | - | 9 - 10 | 2278 | 70 |
AF124726.1 | - | 9 - 10 | 2768 | 70 | |
AF124727.1 | - | 3 - 4 | 304 | 70 | |
AF124728.1 | - | 2 - 3 | 279 | 70 | |
AK299034.1 | - | 8 - 9 | 2160 | 70 | |
AL833228.1 | - | 4 - 5 | 622 | 70 | |
BC140805.1 | - | 9 - 10 | 2538 | 70 | |
BX247975.1 | - | 2 - 3 | 558 | 70 | |
CX786860.1 | - | 3 - 4 | 318 | 70 |