The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032826.4 | NP_116215.1 | 3 - 4 | 620 | 137 |
XM_011516645.2 | XP_011514947.1 | 3 - 4 | 388 | 137 | |
GenBank mRNA | AB052892.1 | - | 3 - 4 | 596 | 137 |
AJ315497.1 | - | 3 - 4 | 452 | 137 | |
AJ315498.1 | - | 3 - 4 | 442 | 137 | |
AJ971941.1 | - | 3 - 4 | 296 | 137 | |
AK027603.1 | - | 3 - 4 | 460 | 137 | |
AK027726.1 | - | 2 - 3 | 178 | 137 | |
AK075368.1 | - | 3 - 4 | 425 | 137 | |
BC008413.1 | - | 3 - 4 | 441 | 137 |