Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35B4
Entrez Gene ID
84912
Gene Name
solute carrier family 35 member B4
Gene Aliases
YEA, YEA4
Chromosome Location
Chr.7: 134289337 - 134320307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032826.4 NP_116215.1 9 - 10 1079 97
XM_011516645.2 XP_011514947.1 9 - 10 847 97
GenBank mRNA AB052892.1 - 9 - 10 1055 97
AJ315497.1 - 9 - 10 911 97
AJ971941.1 - 9 - 10 755 97
AK027603.1 - 9 - 10 919 97
AK075368.1 - 9 - 10 884 97
AK222874.1 - 9 - 10 928 97
AK307621.1 - 8 - 9 857 97
BC008413.1 - 9 - 10 900 97

View Full Product Details