Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC9C
Entrez Gene ID
283237
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
-
Chromosome Location
Chr.11: 62728073 - 62738636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318813.1 NP_001305742.1 1 - 2 258 147
NM_001318814.1 NP_001305743.1 1 - 2 258 147
NM_001318815.1 NP_001305744.1 1 - 2 258 147
GenBank mRNA AF289605.1 - 1 - 2 627 147
BQ050575.1 - 1 - 2 223 147
BX421356.2 - 1 - 2 330 146
CN430374.1 - 1 - 2 196 147

View Full Product Details