Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXF1
Entrez Gene ID
10482
Gene Name
nuclear RNA export factor 1
Gene Aliases
MEX67, TAP
Chromosome Location
Chr.11: 62792125 - 62805492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081491.1 NP_001074960.1 11 - 12 3063 100
NM_006362.4 NP_006353.2 12 - 13 1262 100
GenBank mRNA AB209915.1 - 11 - 12 2998 100
AF112880.1 - 12 - 13 1126 100
AF126246.1 - 12 - 13 1126 100
AJ132712.1 - 12 - 13 1126 100
AK027192.1 - 2 - 3 1199 101
AK307998.1 - 12 - 13 1210 100
BC004904.2 - 12 - 13 1199 100
BC028041.1 - 12 - 13 1192 100
CR749674.1 - 2 - 3 903 100
U80073.1 - 11 - 12 946 100

View Full Product Details