The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081491.1 | NP_001074960.1 | 13 - 14 | 3223 | 98 |
NM_006362.4 | NP_006353.2 | 14 - 15 | 1422 | 98 | |
GenBank mRNA | AB209915.1 | - | 13 - 14 | 3158 | 98 |
AF112880.1 | - | 14 - 15 | 1286 | 98 | |
AF126246.1 | - | 14 - 15 | 1286 | 98 | |
AJ132712.1 | - | 14 - 15 | 1286 | 98 | |
AK027192.1 | - | 4 - 5 | 1359 | 98 | |
AK307961.1 | - | 15 - 16 | 1527 | 98 | |
AK307998.1 | - | 14 - 15 | 1370 | 98 | |
BC004904.2 | - | 14 - 15 | 1359 | 98 | |
BC028041.1 | - | 14 - 15 | 1352 | 98 | |
CR749674.1 | - | 4 - 5 | 1063 | 98 | |
U80073.1 | - | 13 - 14 | 1106 | 98 |