The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081491.1 | NP_001074960.1 | 19 - 20 | 3763 | 65 |
NM_006362.4 | NP_006353.2 | 20 - 21 | 1962 | 65 | |
GenBank mRNA | AB209915.1 | - | 19 - 20 | 3698 | 65 |
BC004904.2 | - | 20 - 21 | 1899 | 65 | |
BC028041.1 | - | 20 - 21 | 1892 | 65 | |
CR749674.1 | - | 10 - 11 | 1603 | 65 |