The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005420.2 | NP_005411.1 | 4 - 5 | 472 | 79 |
XM_011532210.2 | XP_011530512.1 | 4 - 5 | 471 | 79 | |
GenBank mRNA | AK310845.1 | - | 4 - 5 | 465 | 79 |
AK312236.1 | - | 3 - 4 | 371 | 79 | |
BC027956.1 | - | 4 - 5 | 472 | 79 | |
CR407621.1 | - | 3 - 4 | 359 | 79 | |
S77383.1 | - | 4 - 5 | 395 | 79 | |
U08098.1 | - | 4 - 5 | 465 | 79 | |
Y11195.1 | - | 4 - 5 | 421 | 79 |