The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 13 - 14 | 1729 | 115 |
XM_005273833.4 | XP_005273890.1 | 13 - 14 | 2851 | 115 | |
XM_005273834.1 | XP_005273891.1 | 12 - 13 | 1641 | 115 | |
XM_011544811.2 | XP_011543113.1 | 9 - 10 | 1064 | 115 | |
XM_017017339.1 | XP_016872828.1 | 14 - 15 | 1805 | 115 | |
GenBank mRNA | AK094193.1 | - | 10 - 11 | 1240 | 115 |
AK127649.1 | - | 2 - 3 | 171 | 115 | |
AK127943.1 | - | 12 - 13 | 1599 | 115 | |
AK293213.1 | - | 8 - 9 | 922 | 115 | |
AL831992.2 | - | 13 - 14 | 1670 | 115 | |
BC065264.1 | - | 10 - 11 | 1223 | 115 | |
BC109038.1 | - | 10 - 11 | 1184 | 115 | |
BC109039.1 | - | 10 - 11 | 1184 | 115 | |
BC111047.1 | - | 11 - 12 | 1391 | 115 |