The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 5 - 6 | 760 | 99 |
XM_005273833.4 | XP_005273890.1 | 5 - 6 | 1882 | 99 | |
XM_005273834.1 | XP_005273891.1 | 5 - 6 | 762 | 99 | |
GenBank mRNA | AK094193.1 | - | 2 - 3 | 271 | 99 |
AK127943.1 | - | 5 - 6 | 720 | 99 | |
AL831992.2 | - | 5 - 6 | 701 | 99 | |
BC065264.1 | - | 2 - 3 | 254 | 99 | |
BC109038.1 | - | 2 - 3 | 215 | 99 | |
BC109039.1 | - | 2 - 3 | 215 | 99 | |
BC111047.1 | - | 3 - 4 | 422 | 99 |