The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198514.3 | NP_940916.2 | 4 - 5 | 1153 | 105 |
XM_011539769.2 | XP_011538071.1 | 4 - 5 | 1153 | 105 | |
GenBank mRNA | AK090631.1 | - | 4 - 5 | 1124 | 105 |
AK098487.1 | - | 2 - 3 | 267 | 105 | |
BX647641.1 | - | 2 - 3 | 524 | 105 |