The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198514.3 | NP_940916.2 | 10 - 11 | 2165 | 97 |
GenBank mRNA | AK098487.1 | - | 8 - 9 | 1279 | 97 |
AL832090.1 | - | 1 - 2 | 136 | 97 | |
BC032598.1 | - | 6 - 7 | 929 | 97 | |
BX538287.1 | - | 6 - 7 | 959 | 97 | |
BX647641.1 | - | 8 - 9 | 1536 | 97 |