Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
Chromosome Location
Chr.15: 25672241 - 25865144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024490.3 NP_077816.1 1 - 2 555 96
XM_005268261.4 XP_005268318.1 2 - 3 685 96
XM_011521826.2 XP_011520128.1 2 - 3 856 96
XM_011521828.2 XP_011520130.1 1 - 2 1284 96
GenBank mRNA AB051358.1 - 2 - 3 648 96
BC052251.1 - 1 - 2 561 96

View Full Product Details