Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TINAG
Entrez Gene ID
27283
Gene Name
tubulointerstitial nephritis antigen
Gene Aliases
TIN-AG
Chromosome Location
Chr.6: 54307792 - 54390152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014464.3 NP_055279.3 4 - 5 762 70
XM_006715062.2 XP_006715125.1 5 - 6 745 70
XM_011514497.2 XP_011512799.1 2 - 3 594 70
XM_017010745.1 XP_016866234.1 4 - 5 726 70
XM_017010746.1 XP_016866235.1 4 - 5 520 70
XM_017010747.1 XP_016866236.1 2 - 3 371 70
XM_017010748.1 XP_016866237.1 2 - 3 200 70
GenBank mRNA AF195116.1 - 4 - 5 616 70
AK312918.1 - 4 - 5 726 70

View Full Product Details