The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014464.3 | NP_055279.3 | 6 - 7 | 1046 | 75 |
XM_006715062.2 | XP_006715125.1 | 7 - 8 | 1029 | 75 | |
XM_011514497.2 | XP_011512799.1 | 4 - 5 | 878 | 75 | |
XM_017010745.1 | XP_016866234.1 | 6 - 7 | 1010 | 75 | |
XM_017010746.1 | XP_016866235.1 | 6 - 7 | 804 | 75 | |
XM_017010747.1 | XP_016866236.1 | 4 - 5 | 655 | 75 | |
XM_017010748.1 | XP_016866237.1 | 4 - 5 | 484 | 75 | |
GenBank mRNA | AB022277.1 | - | 6 - 7 | 901 | 75 |
AF195116.1 | - | 6 - 7 | 900 | 75 | |
AK312918.1 | - | 6 - 7 | 1010 | 75 | |
BC070278.1 | - | 6 - 7 | 1046 | 75 |