Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TINAG
Entrez Gene ID
27283
Gene Name
tubulointerstitial nephritis antigen
Gene Aliases
TIN-AG
Chromosome Location
Chr.6: 54307792 - 54390152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014464.3 NP_055279.3 7 - 8 1228 119
XM_006715062.2 XP_006715125.1 8 - 9 1211 119
XM_011514497.2 XP_011512799.1 5 - 6 1060 119
XM_017010745.1 XP_016866234.1 7 - 8 1192 119
XM_017010746.1 XP_016866235.1 7 - 8 986 119
XM_017010747.1 XP_016866236.1 5 - 6 837 119
XM_017010748.1 XP_016866237.1 5 - 6 666 119
GenBank mRNA AB022277.1 - 7 - 8 1083 119
AF195116.1 - 7 - 8 1082 119
AF195117.1 - 3 - 4 653 119
AK312918.1 - 7 - 8 1192 119
BC070278.1 - 7 - 8 1228 119

View Full Product Details